Assay Details
Amplicon Length: | 70 |
Chromosome Location: | Chr. 6 - 133884395 - 133884395 [-] on 133884395 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-134-3p | MIMAT0017125 |
| Rat | rno-miR-134-5p | MIMAT0000840 |
| Rat | rno-miR-382-5p | MIMAT0003201 |
| Rat | rno-miR-382-3p | MIMAT0003202 |
Stem-loop Details
Rat

Genomic Map