Assay Details
Gene Family ID: | MIPF0000112, mir-134 |
Mature miRNA Sequence: | UGUGACUGGUUGACCAGAGGGG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-134-5p | MIMAT0000447 | |
| Mouse | mmu-miR-134-5p | MIMAT0000146 | |
| Rat | rno-miR-134-5p | MIMAT0000840 | |
| Callithrix jacchus | cja-miR-134 | MIMAT0039508 | |
| Cricetulus griseus | cgr-miR-134 | MIMAT0023760 | |
| Dog | cfa-miR-134 | MIMAT0009883 | |
| View More Rows | |||
| Goat | chi-miR-134 | MIMAT0035951 | |
| Guinea pig | cpo-miR-134-5p | MIMAT0046990 | |
| Horse | eca-miR-134 | MIMAT0013127 | |
| Pan troglodytes | ptr-miR-134 | MIMAT0008031 | |
| Rhesus monkey | mml-miR-134-5p | MIMAT0006190 | |
Stem-loop Details
Human
Mouse
Rat
Callithrix jacchus
Cricetulus griseus
Dog
Goat
Guinea pig
Horse
Pan troglodytes
Rhesus monkey
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Human Pool AMegaplex™ RT Primers, Rodent Pool A
Related Products
miRBase Alias:
Human: hsa-miR-134 (v19)
Mouse: mmu-miR-134 (v17)
Rat: rno-miR-134 (v18)
Macaca mulatta: mml-miR-134 (v19)
Mouse: mmu-miR-134 (v17)
Rat: rno-miR-134 (v18)
Macaca mulatta: mml-miR-134 (v19)

Genomic Map