Assay Details
Gene Family ID: | MIPF0000112, mir-134 |
Mature miRNA Sequence: | UGUGACUGGUUGACCAGAGGGG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-134-5p | MIMAT0000447 |
| Mouse | mmu-miR-134-5p | MIMAT0000146 |
| Rat | rno-miR-134-5p | MIMAT0000840 |
| Callithrix jacchus | cja-miR-134 | MIMAT0039508 |
| Cricetulus griseus | cgr-miR-134 | MIMAT0023760 |
| Dog | cfa-miR-134 | MIMAT0009883 |
| View More Rows | ||
| Goat | chi-miR-134 | MIMAT0035951 |
| Guinea pig | cpo-miR-134-5p | MIMAT0046990 |
| Horse | eca-miR-134 | MIMAT0013127 |
| Pan troglodytes | ptr-miR-134 | MIMAT0008031 |
| Rhesus monkey | mml-miR-134-5p | MIMAT0006190 |
Stem-loop Details
Human
Mouse
Rat
Callithrix jacchus
Cricetulus griseus
Dog
Goat
Guinea pig
Horse
Pan troglodytes
Rhesus monkey

Genomic Map