Learn More
Thermo Scientific™ Single nanoViper™ and Double nanoViper™ PepMap™ Neo UHPLC Columns
Description
Separate challenging peptide mapping samples with Thermo Scientific™ Single nanoViper™ and Double nanoViper™ PepMap™ Neo UHPLC columns. These columns feature flexible connectivity, high reproducibilty, and excellent separation performance. Our PepMap Neo columns are packed to higher pressure and provide 1500 bar pressure capability, improved column-to-column consistency, and increased efficiency. The column media is manufactured and selected to exacting standards and packed at high pressure, resulting in enhanced peak symmetry, resolution, and column-to-column reproducibility that allows you to reliably obtain greater sample coverage.
Benefits of PepMap Neo Columns
- Brilliant Connectivity — Double nanoViper™ columns connect with ease.
- Brilliant Reproducibility — Precise manufacturing protocols for packing media improve column-to-column consistency for reproducible analysis every day.
- Brilliant Separation — Ultra-high pressure packing increases column efficiency, allowing you to run at up to 1500 bar for enhanced peak symmetry and resolution, providing higher sample capacity for more sample insights.
Specifications
Specifications
| Column Format | Analytical Column, Double nanoViper™ |
| Diameter (Metric) | 75 μm |
| Length (Metric) | 750 mm |
| Max. Pressure | 22,000 psi (1500 bar) |
| Particle Size | 2 μm |
| pH | 2 to 8 |
| Pore Size | 100 Å |
| Quantity | Each |
| Temperature | 60°C |
| USP Type | L1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.