Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Whatman™ Mini-UniPrep™ Syringeless Filters

Catalog No. 09923100 Shop All Cytiva Products
Change view
Click to view available options
Pore Size:
0.2 μm
0.45 μm
Material (Membrane):
Glass Microfiber
Nylon
PES
PTFE
PVDF
Polypropylene
Polypropylene Depth Filter
Quantity:
100/Pk.
1000/Pk.
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size Material (Membrane) Quantity
09-923-100 0.2 μm PVDF 100/Pk.
09-923-25 0.45 μm PVDF 100/Pk.
09-923-26 0.45 μm Polypropylene Depth Filter 100/Pk.
09-923-36 0.45 μm Nylon 1000/Pk.
09-923-28 0.45 μm PTFE 100/Pk.
09-923-29 0.45 μm Nylon 100/Pk.
09-923-106 0.2 μm Nylon 1000/Pk.
09-924-586 0.45 μm Polypropylene Depth Filter 1000/Pk.
09-923-101 0.2 μm Nylon 100/Pk.
09-923-32 0.45 μm PVDF 1000/Pk.
09-923-35 0.45 μm PTFE 1000/Pk.
09-923-27 0.45 μm Glass Microfiber 100/Pk.
09-923-102 0.2 μm PTFE 100/Pk.
09-923-105 0.2 μm PVDF 1000/Pk.
09-923-31 0.45 μm Polypropylene -
09-923-30 0.45 μm PES 100/Pk.
09-924-587 0.45 μm PES 1000/Pk.
09-923-38 0.45 μm Polypropylene -
09-923-34 0.45 μm Glass Microfiber 1000/Pk.
09-927-651 0.2 μm PES 100/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
Catalog No. 09-923-100 Supplier Cytiva Supplier No. UN203NPEAQU
Add to Cart
Add to Cart

Processes sample loads in one-third the time. Whatman™ Mini-UniPrep™ Syringeless Filters provide a faster and easier way to remove particulates from samples being prepared for HPLC and UHPLC analysis.

Whatman Mini-UniPrep™ Transparent Syringeless Filters quickly and easily remove particulates from samples being prepared for HPLC/UHPLC analysis, while maintaining sample visibility.

  • Pre-assembled all-in-one filtration unit eliminates the need for syringe filter, syringe, vial and septum, reducing costs by up to 40%
  • Wide range of membrane choices in 0.2 and 0.45 μm pore sizes to meet specific sample application requirements
  • Compatible with most major autosamplers for high-throughput analysis
  • Fewer consumables required
  • Plunger contains a filtration membrane at one end and a pre-attached cap/septum at the other
  • Can be used either manually or with a compressor unit
  • Multi-sample compressor processes up to 6 samples at a time, further improving sample processing time and reducing hand stress
  • Designed to fit into any autosampler accommodating 12 x 32mm vials -OR-
    Septum can be pierced with needle and sample drawn off for manual injection into an analyzer
TRUSTED_SUSTAINABILITY

Specifications

Color Translucent
Disposable Single-use
For Use With (Equipment) Autosampler, Compressor
Material Septa: PTFE-coated Silicone Rubber
Material (Membrane) PVDF
Pore Size 0.2 μm
For Use With (Application) Routine HPLC/UHPLC analysis; Sample filtration; Composite assays; Content uniformity; Protein precipitation; Solubility/dissolubility testing; Ion chromatography
Housing Material Polypropylene (PP)
Includes Vial with incorporated filtration membrane, plunger and cap/septa
Pressure Rating 0.6 bar / 8 psi (to compress)
Quantity 100/Pk.
Temperature (Metric) Max 50°C
Type Syringeless Filter
Volume (Metric) 0.4 mL
Height (Metric) 32 mm
Diameter (Metric) 12 mm
Closure Type Cap with Septa
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.