Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,1-Trichloro-2-methyl-2-propanol hemihydrate, 98%

Catalog No. AC150300025
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle

CAS: 6001-64-5 | 0·5 H2O | 186.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6001-64-5
Molecular Formula 0·5 H2O
Molecular Weight (g/mol) 186.47
MDL Number MFCD02179352
InChI Key HBARVHNVVKZUGS-UHFFFAOYSA-N
Synonym c4h7cl3o.1/2h2o, 1,1,1-trichloro-2-methyl-2-propanol hemihydrate, chlorobutanol hydrogen ion hydrate
PubChem CID 102594540
IUPAC Name molecular hydrogen;1,1,1-trichloro-2-methylpropan-2-ol;hydrate
SMILES [HH].CC(C)(C(Cl)(Cl)Cl)O.O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 75°C to 79°C
Color White
Boiling Point 173°C to 175°C
Flash Point 100°C
Infrared Spectrum Authentic
Assay Percent Range 97.5 to 102.5% (ex Cl) (Argentometry)
Packaging Plastic bottle
Linear Formula 0·5H2O
Quantity 2.5 kg
Solubility Information Solubility in water: 7.7g/L (20°C). Other solubilities: soluble in ethanol,ether,chloroform,glycerol
Formula Weight 186.47
Percent Purity 98%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 1, 1, 1-Trichloro-2-methyl-2-propanol hemihydrate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.