Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,2,2-Tetrachloroethane, 98.5%

Catalog No. AC147940010
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass bottle

CAS: 79-34-5 | C2H2Cl4 | 167.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-34-5
MDL Number MFCD00000848
InChI Key QPFMBZIOSGYJDE-UHFFFAOYSA-N
Synonym s-tetrachloroethane, acetylene tetrachloride, ethane, 1,1,2,2-tetrachloro, bonoform, cellon, tetrachlorethane, sym-tetrachloroethane, westron, 1,1,2,2-tetrachlorethane, 1,1,2,2-tetrachloraethan
PubChem CID 6591
ChEBI CHEBI:36026
IUPAC Name 1,1,2,2-tetrachloroethane
SMILES C(C(Cl)Cl)(Cl)Cl

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -43.0°C
Color Colorless
Density 1.5800g/mL
Boiling Point 147.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5%
Packaging Glass bottle
Molecular Formula C2H2Cl4
Refractive Index 1.4930 to 1.4950
Linear Formula Cl2CHCHCl2
Quantity 1 L
Beilstein 01, 86
Specific Gravity 1.58
Merck Index 15, 9334
Solubility Information Solubility in water: 0.3 g/100mL (25°C). Other solubilities: miscible with methanol, ethanol, benzene, ether,, petroleum ether, carbon tetrachloride, chloroform,, carbon disulfide, dimethylformamide and oils, soluble in acetone
Molecular Weight (g/mol) 167.85
Viscosity 1.7 mPa.s (28°C)
Formula Weight 167.85
Percent Purity 98.5%
Physical Form Liquid
Chemical Name or Material 1,1,2,2-Tetrachloroethane, 98.5%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if inhaled.
Fatal in contact with skin.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTE
EINECSNumber 201-197-8
RTECSNumber KI8575000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.