Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1'-(2,6-Pyridinediyl)bis(3-methylimidazolium) Dibromide 98.0+%, TCI America™
SDP

Catalog No. p18165g Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18165G 5 g
P18161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P18165G Supplier TCI America Supplier No. P18165G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 263874-05-1
Molecular Formula C13H15Br2N5
Molecular Weight (g/mol) 401.11
MDL Number MFCD11975436
InChI Key VAESRVWVZJLVIA-UHFFFAOYSA-L
PubChem CID 11235002
IUPAC Name 3-methyl-1-[6-(3-methyl-1H-imidazol-3-ium-1-yl)pyridin-2-yl]-1H-imidazol-3-ium dibromide
SMILES [Br-].[Br-].C[N+]1=CN(C=C1)C1=CC=CC(=N1)N1C=C[N+](C)=C1

Specifications

Quantity 5 g
Formula Weight 401.11
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-(2,6-Pyridinediyl)bis(3-methylimidazolium) Dibromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.