Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,10-Phenanthroline Monohydrate, 0.1% w/v, Reagents

Catalog No. 01334282
Change view
Click to view available options
:
Quantity: 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-334-282 Quantity: 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-334-282 Supplier Reagents Holdings Llc Supplier No. CP1219001C

Due to product restrictions, please to purchase or view availability for this product.

1,10-Phenanthroline Monohydrate, 0.1% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 5144-89-8
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym o-Phenanthroline
IUPAC Name water
SMILES O

Specifications

Color Colorless
Boiling Point 100°C
Packaging Glass Bottle
Quantity 1 L
Specific Gravity 1.001
Percent Purity 0.1%
Grade Reagent
Physical Form Liquid
Chemical Name or Material 1,10-Phenanthroline Monohydrate, 0.1% w/v
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.