Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3,4-Tetrahydroisoquinoline-1-acetic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
105400-81-5
C11H13NO2
191.23
MFCD00215024
BUAVPRGEIAVFBF-UHFFFAOYSA-N
1,2,3,4-tetrahydroisoquinoline-1-yl acetic acid,2-1,2,3,4-tetrahydroisoquinolin-1-yl acetic acid,1,2,3,4-tetrahydroisoquinoline-1-acetic acid,1,2,3,4-tetrahydro-isoquinolin-1-yl-acetic acid,2-1,2,3,4-tetrahydro-1-isoquinolinyl acetic acid,1,2,3,4-tetrahydroisoquinolin-1-ylacetic acid,1-isoquinolineaceticacid, 1,2,3,4-tetrahydro,1,2,3,4-tetrahydro-1-isoquinolinylacetic acid,2-1,2,3,4-tetrahydroisoquinolyl acetic acid,tetrahydroisoquinolylacetic acid
2733683
2-(1,2,3,4-tetrahydroisoquinolin-1-yl)acetic acid
C1CNC(C2=CC=CC=C21)CC(=O)O
105400-81-5
C11H13NO2
191.23
MFCD00215024
BUAVPRGEIAVFBF-UHFFFAOYSA-N
1,2,3,4-tetrahydroisoquinoline-1-yl acetic acid,2-1,2,3,4-tetrahydroisoquinolin-1-yl acetic acid,1,2,3,4-tetrahydroisoquinoline-1-acetic acid,1,2,3,4-tetrahydro-isoquinolin-1-yl-acetic acid,2-1,2,3,4-tetrahydro-1-isoquinolinyl acetic acid,1,2,3,4-tetrahydroisoquinolin-1-ylacetic acid,1-isoquinolineaceticacid, 1,2,3,4-tetrahydro,1,2,3,4-tetrahydro-1-isoquinolinylacetic acid,2-1,2,3,4-tetrahydroisoquinolyl acetic acid,tetrahydroisoquinolylacetic acid
2733683
2-(1,2,3,4-tetrahydroisoquinolin-1-yl)acetic acid
C1CNC(C2=CC=CC=C21)CC(=O)O
Quantity 1g
Melting Point 248.0°C to 251.0°C
Packaging Glass bottle
Color White
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in methanol
Formula Weight 191.23
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 1,2,3,4-Tetrahydroisoquinoline-1-acetic acid, 98%
Show More Show Less