Learn More
1,2,4-Trichlorobenzene, Spectrophotometric Grade, 99% min
Description
1,2,4-Trichlorobenzene is used as a dielectric and heat transfer fluid in transformers. It acts as an intermediate, degreaser, wood preservative and solvent for dye. It is a high-temperature solvent used in gel permeation chromatography, especially for polyethylene and polypropylene. Further, it is used as a lubricant and as a synthetic transformer oil.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 17°C |
| Color | Colorless |
| Density | 1.454g/cm3 at 20°C |
| Boiling Point | 213°C |
| Quantity | 4 L |
| Flash Point | 110°C (230°F) |
| Ignition Point | 571°C (1060°F) |
| Odor | Aromatic |
| Physical Form | Liquid |
| Refractive Index | 1.572 |
| Show More |
Safety and Handling
| Hazard Category | H302-H315-H319 |
| Precautionary Statement | P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c |
| DOTInformation | DOT Class: 6.1, Packaging Group: III |
| EINECSNumber | 204-428-0 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.