Alfa Aesar™ 1-(2,5-Difluorobenzyl)piperazine, 97%

Manufacturer: Alfa Aesar™ H5502103

Quantity 1g

 View more versions of this product

Catalog No. AAH5502103

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 179334-18-0
    Molecular Formula C11H14F2N2
    Formula Weight 212.24
    MDL Number MFCD05189212
    Chemical Name or Material 1-(2,5-Difluorobenzyl)piperazine

    Specifications

    Formula Weight 212.24
    MDL Number MFCD05189212
    Flash Point >110°C (230°F)
    Molecular Formula C11H14F2N2
    CAS 179334-18-0
    Boiling Point 80° to 82°C (0.04mmHg)
    Quantity 1g
    Chemical Name or Material 1-(2,5-Difluorobenzyl)piperazine
    Assay 97%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit