Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Bis(2-chloroethoxy)ethane, 97%

Catalog No. p-7026717
Click to view available options
Quantity:
100 g
500 g

CAS: 112-26-5 | C6H12Cl2O2 | 187.06 g/mol

1,2-Bis(2-chloroethoxy)ethane is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Bis(2-chloroethoxy)ethane is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-26-5
Molecular Formula C6H12Cl2O2
Molecular Weight (g/mol) 187.06
MDL Number MFCD00000976
InChI Key AGYUOJIYYGGHKV-UHFFFAOYSA-N
Synonym 1,2-bis 2-chloroethoxy ethane, triglycol dichloride, triethylene glycol dichloride, ethane, 1,2-bis 2-chloroethoxy, 1,8-dichloro-3,6-dioxaoctane, 1,2-bis chloroethoxy ethane, bis 2-chloroethoxy ethane, di 2-chloroethyl cellosolve, 2-2-chloroethoxy ethyl 2'-chloroethyl ether, unii-28jq82m13j
PubChem CID 8171
IUPAC Name 1,2-bis(2-chloroethoxy)ethane
SMILES ClCCOCCOCCCl

Specifications

Melting Point -32°C
Density 1.197
Boiling Point 235°C
Flash Point >110°C (230°F)
Refractive Index 1.461
Quantity 100 g
UN Number UN2810
Beilstein 506177
Solubility Information Insoluble in water.
Formula Weight 187.07
Percent Purity 97%
Chemical Name or Material 1,2-Bis(2-chloroethoxy)ethane
Show More Show Less
Hazard Category H302-H312-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H315-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 203-952-7
RTECSNumber KH4900000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.