Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1,2-Dichlorobenzene, Spectrophotometric Grade, 98%
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
Description
1,2-Dichlorobenzene is used in spectrophotometry and environmental testing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | °C to 18°C |
| Color | Colorless |
| Density | 1.306 g/cm3 at 20°C |
| Boiling Point | 180°C |
| Flash Point | 68°C (154°F) |
| Ignition Point | 640°C (1184°F) |
| Odor | Pungent |
| Assay Percent Range | 98% |
| Refractive Index | 1.551 |
| Quantity | 4 L |
| Show More |
Safety and Handling
| Hazard Category | H227-H302+H332-H315-H317-H319-H335 |
| Hazard Statement | This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets. |
| Precautionary Statement | P210-P235-P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P370+P378q-P501c |
| DOTInformation | DOT Class: 6.1, Packaging Group: III |
| EINECSNumber | 202-425-9 |
| RTECSNumber | CZ4500000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction