Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dichlorobenzene, 99+%, for HPLC, Acros Organics

Click to view available options
Quantity:
100mL
1L
Packaging:
Glass Bottle
95-50-1
C6H4Cl2
147
RFFLAFLAYFXFSW-UHFFFAOYSA-N
o-dichlorobenzene,chloroben,o-dichlorbenzol,dilantin db,ortho-dichlorobenzene,2-dichlorobenzene,o-dichlorobenzol,odcb,benzene, 1,2-dichloro,cloroben
7239
CHEBI:35290
1,2-dichlorobenzene
C1=CC=C(C(=C1)Cl)Cl
95-50-1
C6H4Cl2
147
RFFLAFLAYFXFSW-UHFFFAOYSA-N
o-dichlorobenzene,chloroben,o-dichlorbenzol,dilantin db,ortho-dichlorobenzene,2-dichlorobenzene,o-dichlorobenzol,odcb,benzene, 1,2-dichloro,cloroben
7239
CHEBI:35290
1,2-dichlorobenzene
C1=CC=C(C(=C1)Cl)Cl
Melting Point -15.0°C
Density 1.3000g/mL
Packaging Glass Bottle
Flash Point 67°C
Boiling Point 179.0°C to 180.0°C
Refractive Index 1.5500 to 1.5520
Specific Gravity 1.3
Quantity 1L
Assay Percent Range 99% min. (GC)
Linear Formula C6H4Cl2
Merck Index 15,3065
Solubility Information Solubility in water: 0.13g/L(20°C). Other solubilities: miscible with most common organic solvents,miscible with alcohol,ether and benzene
Formula Weight 147
Percent Purity 99+%
Grade HPLC
Chemical Name or Material 1, 2-Dichlorobenzene
Show More Show Less

GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.
Causes skin irritation.
Harmful if inhaled.
May cause

GHS P Statement
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritatio

GHS Signal Word: Warning

EINECSNumber : 202-425-9