Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1,2-Dichlorobenzene, 99%
Click to view available options
Quantity:
10,000 g
2500 g
500 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications
1,2-Dichlorobenzene is widely used as a precursor to prepare 1,2-dichloro-4-nitrobenzene, which is an intermediate in the synthesis of agrochemicals. It acts as a solvent for fullerenes. It is also used in softening and removing carbon-based contamination on metal surfaces. It also used as food preservative, motor oil additive, lubricant and paint.
Solubility
Miscible with water and most of the organic solvents.
Notes
Keep away from sources of ignition. Incompatible with strong oxidizing agents.
1,2-Dichlorobenzene is widely used as a precursor to prepare 1,2-dichloro-4-nitrobenzene, which is an intermediate in the synthesis of agrochemicals. It acts as a solvent for fullerenes. It is also used in softening and removing carbon-based contamination on metal surfaces. It also used as food preservative, motor oil additive, lubricant and paint.
Solubility
Miscible with water and most of the organic solvents.
Notes
Keep away from sources of ignition. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | -18°C |
| Density | 1.306 |
| Boiling Point | 180°C |
| Flash Point | 68°C (154°F) |
| Odor | Pungent |
| Refractive Index | 1.551 |
| Quantity | 10,000 g |
| UN Number | UN1591 |
| Beilstein | 606078 |
| Merck Index | 14,3056 |
| Show More |
Safety and Handling
| Hazard Category | H227-H302+H332-H315-H317-H319-H335 |
| Hazard Statement | GHS H Statement H302-H315-H319-H335-H227 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Combustible liquid. |
| Precautionary Statement | P210-P235-P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: O-DICHLOROBENZENE |
| EINECSNumber | 202-425-9 |
| RTECSNumber | CZ4500000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction