Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dimethoxyethane, Reagent, 99.5%, Spectrum™

Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
110-71-4
C4H10O2
90.12
XTHFKEDIFFGKHM-UHFFFAOYSA-N
1,2-dimethoxyethane
COCCOC
110-71-4
C4H10O2
90.12
XTHFKEDIFFGKHM-UHFFFAOYSA-N
1,2-dimethoxyethane
COCCOC

DI139, 110-71-4, C4H10O2

Spectrum™ 1,2-Dimethoxyethane, Reagent is also known as glyme, monoglyme, dimethyl glycol, ethylene glycol dimethyl ether, dimethyl cellosolve and DME. Dimethoxyethane is a clear, colorless, aprotic liquid ether. It is commonly used as a solvent in batteries and in organometallic reaction chemistry since it is higher boiling than diethyl ether and THF. It is also miscible with water. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.
Acidity 0.015%
Water 0.05%
Packaging Amber Glass Bottle
Quantity 100 mL
CAS Max % 100%
UN Number UN2252
Percent Purity 99.5%