Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dimethoxyethane, Reagent, 99.5%, Spectrum™ Chemical
SDP

Catalog No. 18600036 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
This item is not returnable. View return policy
This item is not returnable. View return policy

DI139, 110-71-4, C4H10O2

Spectrum™ Chemical 1,2-Dimethoxyethane, Reagent is also known as glyme, monoglyme, dimethyl glycol, ethylene glycol dimethyl ether, dimethyl cellosolve and DME. Dimethoxyethane is a clear, colorless, aprotic liquid ether. It is commonly used as a solvent in batteries and in organometallic reaction chemistry since it is higher boiling than diethyl ether and THF. It is also miscible with water. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 110-71-4
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
InChI Key XTHFKEDIFFGKHM-UHFFFAOYSA-N
IUPAC Name 1,2-dimethoxyethane
SMILES COCCOC

Specifications

Acidity 0.015%
CAS Max % 100%
Packaging Steel Can
Quantity 20 L
UN Number UN2252
Percent Purity 99.5%
Grade Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.