Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-(2'-Fluoro[1,1'-biphenyl]-4-yl)propan-1-one, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Specifications
Specifications
| CAS Min % | 97.0 |
| Assay Percent Range | 97% |
| Packaging | Amber glass bottle |
| Linear Formula | FC6H4C6H4C(O)CH2CH3 |
| Quantity | 1 g |
| Formula Weight | 228.26 |
| Percent Purity | 97% |
| Chemical Name or Material | 1-(2'-Fluoro[1, 1'-biphenyl]-4-yl)propan-1-one |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction