Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bismuth(III) oxyfluoride, 97%, pure, Thermo Scientific™
Click to view available options
Quantity:
25 g
100 g
Specifications
Specifications
| Melting Point | 500.0°C |
| Quantity | 25 g |
| Assay Percent Range | 96% min. (ex Bi) |
| Linear Formula | FBi=O |
| Solubility Information | Solubility in water: insoluble. |
| Formula Weight | 243.98 |
| Percent Purity | 97% |
| Grade | Pure |
| Chemical Name or Material | Bismuth(III) oxyfluoride |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement May be corrosive to metals. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Keep only in |
| EINECSNumber | 236-861-6 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction