Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,5-Triformylbenzene, 98%

Catalog No. p-4380115
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Catalog No. AC446550010

Supplier: Thermo Scientific Chemicals 446550010

Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 3163-76-6 | C9H6O3 | 162.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3163-76-6
Molecular Formula C9H6O3
Molecular Weight (g/mol) 162.14
MDL Number MFCD00182470
InChI Key AEKQNAANFVOBCU-UHFFFAOYSA-N
Synonym 1,3,5-benzenetricarboxaldehyde, 1,3,5-triformylbenzene, trimesaldehyde, 1,3,5-benzenetricarbaldehyde, benzene-1,3,5-tricarboxaldehyde, trimesic formaldehyde, 1,3,5-triformyl benzene, bzoh31, aekqnaanfvobcu-uhfffaoysa
PubChem CID 2747968
IUPAC Name benzene-1,3,5-tricarbaldehyde
SMILES O=CC1=CC(C=O)=CC(C=O)=C1

Specifications

Melting Point 156.0°C to 158.0°C
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Formula Weight 162.14
Percent Purity 98%
Chemical Name or Material 1, 3, 5-Triformylbenzene
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.