Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Bis(tosyloxy)-2,2-dimethylpropane 98.0+%, TCI America™
SDP

Catalog No. B187025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B187025G 25 g
B18705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B187025G Supplier TCI America Supplier No. B187025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 22308-12-9
Molecular Formula C19H24O6S2
Molecular Weight (g/mol) 412.515
MDL Number MFCD00110766
InChI Key LJISGCCLDDOYIJ-UHFFFAOYSA-N
Synonym 1,3-Bis(p-toluenesulfonyloxy)-2,2-dimethylpropane, 2,2-Dimethyl-1,3-propanediol Ditosylate, Neopentyl Glycol Ditosylate
PubChem CID 264253
IUPAC Name [2,2-dimethyl-3-(4-methylphenyl)sulfonyloxypropyl] 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCC(C)(C)COS(=O)(=O)C2=CC=C(C=C2)C

Specifications

Melting Point 121°C
Color White
Quantity 25 g
Formula Weight 412.52
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis(tosyloxy)-2,2-dimethylpropane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.