Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3-Chloro-5-trifluoromethyl-2-pyridyl)homopiperazine, 98%

Catalog No. AAH5026303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5026303 1 g
AAH5026306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5026303 Supplier Thermo Scientific Chemicals Supplier No. H5026303
Only null left
Add to Cart

CAS: 231953-40-5 | C11H13ClF3N3 | 279.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 231953-40-5
Molecular Formula C11H13ClF3N3
Molecular Weight (g/mol) 279.69
MDL Number MFCD00238856
InChI Key PKKLERQDFWUWNM-UHFFFAOYSA-N
Synonym 1-3-chloro-5-trifluoromethyl pyridin-2-yl-1,4-diazepane, 1-3-chloro-5-trifluoromethyl-2-pyridyl-1,4-diazepane, 1-3-chloro-5-trifluoromethyl-2-pyridyl homopiperazine, 1-3-chloro-5-trifluoromethyl pyrid-2-yl homopiperazine, 1-3-chloranyl-5-trifluoromethyl pyridin-2-yl-1,4-diazepane, acmc-20ap02, 1-3-chloro-5-trifluoromethyl pyridin-2-yl homopiperazine, 1-3-chloro-5-trifluoromethyl-2-pyridinyl-1,4-diazepane
PubChem CID 735734
IUPAC Name 1-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-1,4-diazepane
SMILES FC(F)(F)C1=CC(Cl)=C(N=C1)N1CCCNCC1

Specifications

Boiling Point 128°C (1 mmHg)
Quantity 1 g
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 279.69
Percent Purity 98%
Chemical Name or Material 1-(3-Chloro-5-trifluoromethyl-2-pyridyl)homopiperazine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.