Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,3-Dibromo-2-chloro-5-fluorobenzene, 98%

Click to view available options
Quantity:
5g
25g
179897-90-6
C6H2Br2ClF
288.338
MFCD00061122
PZKDJJMHRYNBOR-UHFFFAOYSA-N
1-chloro-2,6-dibromo-4-fluorobenzene,2,6-dibromo-4-fluorochlorobenzene,2-chloro-1,3-dibromo-5-fluorobenzene,4-chloro-3,5-dibromo-1-fluorobenzene,benzene, 1,3-dibromo-2-chloro-5-fluoro,pubchem3411,acmc-1c9lw,2,6-dibromo-4-fluoro-1-chlorobenzene,benzene,1,3-dibromo-2-chloro-5-fluoro
2724604
1,3-dibromo-2-chloro-5-fluorobenzene
C1=C(C=C(C(=C1Br)Cl)Br)F
179897-90-6
C6H2Br2ClF
288.338
MFCD00061122
PZKDJJMHRYNBOR-UHFFFAOYSA-N
1-chloro-2,6-dibromo-4-fluorobenzene,2,6-dibromo-4-fluorochlorobenzene,2-chloro-1,3-dibromo-5-fluorobenzene,4-chloro-3,5-dibromo-1-fluorobenzene,benzene, 1,3-dibromo-2-chloro-5-fluoro,pubchem3411,acmc-1c9lw,2,6-dibromo-4-fluoro-1-chlorobenzene,benzene,1,3-dibromo-2-chloro-5-fluoro
2724604
1,3-dibromo-2-chloro-5-fluorobenzene
C1=C(C=C(C(=C1Br)Cl)Br)F
Quantity 5g
Melting Point 84°C to 87°C
Formula Weight 288.35
Percent Purity 98%
Chemical Name or Material 1,3-Dibromo-2-chloro-5-fluorobenzene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning