Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dibromo-2-propanone, 70%, tech.

Catalog No. ac406550250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC406550250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC406550250 Supplier Thermo Scientific Chemicals Supplier No. 406550250
Only null left
Add to Cart

CAS: 816-39-7 | C3H4Br2O | 215.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 816-39-7
Molecular Formula C3H4Br2O
Molecular Weight (g/mol) 215.87
MDL Number MFCD00013540
InChI Key LQQKDSXCDXHLLF-UHFFFAOYSA-N
Synonym 1,3-dibromoacetone, 1,3-dibromo-2-propanone, 2-propanone, 1,3-dibromo, 1,3-dibromopropanone, bromomethyl ketone, bromomethylketone, sgqdllrxbrwsp@, 1,3-dibromacetone, acmc-20ap4e, 2-propanone,3-dibromo
PubChem CID 69952
IUPAC Name 1,3-dibromopropan-2-one
SMILES C(C(=O)CBr)Br

Specifications

CAS Min % 65.0
CAS Max % 100.0
Color Brown to Yellow
Boiling Point 95°C (20.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 65% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2COCH2Br
Quantity 25 g
Formula Weight 215.87
Percent Purity 70%
Grade Technical
Physical Form Liquid
Chemical Name or Material 1, 3-Dibromo-2-propanone, tech., 70%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 212-430-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.