Learn More
1,3-Dichloropropane, 95%
Description
1,3-Dichloropropane is used as an intermediate in organic synthesis. It is also used as solvent, detergent and chromatographic analysis. Further, it is used to prepare 3,7-dithia-nonanedioic acid by reacting with mercaptoacetic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 1.190 g/mL |
| Boiling Point | 120°C |
| Flash Point | 20°C (68°F) |
| Odor | Sweet |
| Refractive Index | 1.4481 |
| Quantity | 25 g |
| UN Number | UN1993 |
| Solubility Information | Miscible with water. |
| Formula Weight | 112.99 |
| Percent Purity | 95% |
| Show More |
Safety and Handling
| Hazard Category | H225-H302+H332-H315-H319 |
| Hazard Statement | GHS H Statement H225 Highly flammable liquid and vapor. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
| EINECSNumber | 205-531-3 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.