Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dihydroxyacetone dimer, 97%

Catalog No. p-7021733
Click to view available options
Quantity:
50 g
250 g
1000 g

CAS: 62147-49-3 | C3H6O3 | 90.08 g/mol

1,3-Dihydroxyacetone dimer is used as a catalytic agent and petrochemical additive. It is also used as a building block in organic synthesis. Further, it serves as a reagent in three-carbon nucleophilic or electrophilic component in various reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dihydroxyacetone dimer is used as a catalytic agent and petrochemical additive. It is also used as a building block in organic synthesis. Further, it serves as a reagent in three-carbon nucleophilic or electrophilic component in various reactions.

Solubility
Soluble in ethanol, ethyl ether, acetone. Slightly soluble in water. Insoluble in ligroin.

Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62147-49-3
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.08
MDL Number MFCD00051019
InChI Key RXKJFZQQPQGTFL-UHFFFAOYSA-N
Synonym 1,3-dihydroxyacetone dimer, 1,3-dihydroxypropan-2-one dimer, 2,5-bis hydroxymethyl-1,4-dioxane-2,5-diol, 2,5-dihydroxy-1,4-dioxane-2,5-dimethanol, dihydroxyacetone dimer, 2,5-dihydroxydioxane-2,5-dimethanol, 1,4-dioxane-2,5-dimethanol, 2,5-dihydroxy, acmc-20aoav, pubchem4073
PubChem CID 4180364
SMILES OCC(=O)CO

Specifications

Melting Point ∼75°C to 82°C
Quantity 50 g
Beilstein 112910
Merck Index 14,3182
Solubility Information Soluble in ethanol,ethyl ether,acetone. Slightly soluble in water. Insoluble in ligroin.
IUPAC Name 2,5-bis(hydroxymethyl)-1,4-dioxane-2,5-diol
Formula Weight 180.16
Percent Purity 97%
Chemical Name or Material 1,3-Dihydroxyacetone dimer
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 612-947-2
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.