Learn More
1,3-Diphenylguanidine, primary standard, 99+%
Description
Primary material for standardizing acids. Diphenylguanidine is used as a medium of natural and synthetic rubber accelerator. It acts as a complexing agent for the detection of metals and organic bases. Further, it is used as a secondary foam stabilizer in the silicoflouride foam process. It is actively involved in the manufacture of rubber sheet, rubber shoes and other rubber industrial products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 148°C to 150°C |
| Color | Cream |
| Density | 1.130 g/mL |
| Boiling Point | 170°C (decomposition) |
| Odor | Characteristic |
| Quantity | 50 g |
| UN Number | UN3077 |
| Beilstein | 1875653 |
| Merck Index | 14,3324 |
| Solubility Information | Soluble in water,ethanol,carbon tetrachloride,chloroform,toluene and ethyl ether. Slightly soluble in water. |
| Show More |
Safety and Handling
| Hazard Category | H302-H315-H319-H335-H361f |
| Hazard Statement | GHS H Statement H361-H302-H315-H319-H335 Suspected of damaging fertility or the unborn child. Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
| EINECSNumber | 203-002-1 |
| RTECSNumber | MF0875000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.