Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Phenylenediamine-4-sulfonic Acid 98.0+%, TCI America™
SDP

Catalog No. P089325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P089325G 25 g
P0893500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P089325G Supplier TCI America Supplier No. P089325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 88-63-1
Molecular Formula C6H8N2O3S
Molecular Weight (g/mol) 188.201
MDL Number MFCD00035772
InChI Key JVMSQRAXNZPDHF-UHFFFAOYSA-N
Synonym 2,4-Diaminobenzenesulfonic Acid
PubChem CID 66623
IUPAC Name 2,4-diaminobenzenesulfonic acid
SMILES C1=CC(=C(C=C1N)N)S(=O)(=O)O

Specifications

Color Yellow
Quantity 25 g
Formula Weight 188.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Phenylenediamine-4-sulfonic Acid
EINECSNumber (3)-2058
RTECSNumber DB5900000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.