Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Propanesultone, 99%, Thermo Scientific Chemicals

Catalog Number AAA1192309
Click to view available options
Quantity:
10 g
50 g
250 g
1120-71-4
C3H6O3S
122.14
MFCD00005355
FSSPGSAQUIYDCN-UHFFFAOYSA-N
1,3-propanesultone, propane sultone, 1,2-oxathiolane 2,2-dioxide, 1,3-propane sultone, 1,2-oxathiolane, 2,2-dioxide, propanesultone, rcra waste number u193, .gamma.-propane sultone, 3-hydroxy-1-propanesulfonic acid gamma-sultone, unii-l6ntk7vjx9
14264
CHEBI:82370
O=S1(=O)CCCO1
1120-71-4
C3H6O3S
122.14
MFCD00005355
FSSPGSAQUIYDCN-UHFFFAOYSA-N
1,3-propanesultone, propane sultone, 1,2-oxathiolane 2,2-dioxide, 1,3-propane sultone, 1,2-oxathiolane, 2,2-dioxide, propanesultone, rcra waste number u193, .gamma.-propane sultone, 3-hydroxy-1-propanesulfonic acid gamma-sultone, unii-l6ntk7vjx9
14264
CHEBI:82370
O=S1(=O)CCCO1

1,3-Propane sultone is used as a chemical intermediate to introduce sulfopropyl groups into molecules and to confer water solubility and anionic character.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Propane sultone is used as a chemical intermediate to introduce sulfopropyl groups into molecules and to confer water solubility and anionic character.

Solubility
Soluble in water (100 mg/ml), chloroform, esters, methanol, and aromatic hydrocartions.

Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.com
TRUSTED_SUSTAINABILITY
Melting Point 30°C to 33°C
Density 1.392
Boiling Point 179°C to 181°C (30 mm)
Flash Point >110°C (230°F)
Assay Percent Range 99%
Quantity 10 g
Beilstein 109782
Sensitivity Moisture sensitive
Solubility Information Soluble in water (100mg/ml),chloroform,esters,methanol,and aromatic hydrocartions.
IUPAC Name oxathiolane 2,2-dioxide
Formula Weight 122.14
Chemical Name or Material 1, 3-Propanesultone
Show More Show Less
Hazard Category H302+H312-H350
Precautionary Statement P201-P202-P264b-P270-P281-P301+P312-P302+P352-P308+P313-P312-P330-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 214-317-9
RTECSNumber RP5425000
TSCA Yes
storageNote1 Moisture Sensitive
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit