Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3-Sulfopropyl)pyridinium Hydroxide Inner Salt 98.0+%, TCI America™
SDP

Catalog No. S0391250G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
250 g

Catalog No. S0391250G

Supplier: TCI America S0391250G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15471-17-7
Molecular Formula C8H11NO3S
Molecular Weight (g/mol) 201.24
MDL Number MFCD00064468
InChI Key REEBJQTUIJTGAL-UHFFFAOYSA-N
Synonym 3-pyridin-1-ium-1-yl propane-1-sulfonate, ndsb-201, 1-3-sulphonatopropyl pyridinium, unii-qn4i6ai9ek, 3-1-pyridinio-1-propanesulfonate, 1-pyridiniumpropane-3-sulfonate, qn4i6ai9ek, 1-3-sulfopropyl pyridinium hydroxide, inner salt, pyridinium, 1-3-sulfopropyl-, inner salt, 1-3-sulfonatopropyl pyridin-1-ium
PubChem CID 84929
IUPAC Name 1-(3-sulfonatopropyl)pyridin-1-ium
SMILES [O-]S(=O)(=O)CCC[N+]1=CC=CC=C1

Specifications

Melting Point 279°C
Color White
pH 3 to 6 (10% H2O soln. 20°C)
Quantity 250 g
Formula Weight 201.24
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1-(3-Sulfopropyl)pyridinium Hydroxide Inner Salt
EINECSNumber (5)-0758&(5)-3704
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.