Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Bromobenzyl)piperazine, 97%, Thermo Scientific™

Catalog No. p-7052645
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH55145MD 250 mg
AAH5514503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH55145MD Supplier Thermo Scientific Chemicals Supplier No. H55145MD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91345-62-9
Molecular Formula C11H15BrN2
Molecular Weight (g/mol) 255.159
MDL Number MFCD02177416
InChI Key MAHWBNAOEVAPJF-UHFFFAOYSA-N
Synonym 1-4-bromobenzyl piperazine, 1-4-bromophenyl methyl piperazine, 1-4-bromo-benzyl-piperazine, piperazine, 1-4-bromophenyl methyl, cbmicro_035222, acmc-20apb8, cambridge id 5862395, 1-4-bromobenzyl-piperazine, 4-bromophenyl methyl piperazine
PubChem CID 876494
IUPAC Name 1-[(4-bromophenyl)methyl]piperazine
SMILES C1CN(CCN1)CC2=CC=C(C=C2)Br

Specifications

Melting Point 56°C to 62°C
Color Brown to White
Boiling Point 112°C to 116°C (0.3 mmHg)
Quantity 250 mg
UN Number UN3259
Formula Weight 255.16
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(4-Bromobenzyl)piperazine
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.