Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Butanediol diglycidyl ether, 60%, technical, Thermo Scientific Chemicals

Catalog No. p-78053
Click to view available options
Quantity:
500 g
2.5 kg
This item is not returnable. View return policy

Catalog No. AC154315000

Supplier: Thermo Scientific Chemicals 154315000

Only null left
Add to Cart
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2425-79-8
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD00005146
InChI Key SHKUUQIDMUMQQK-UHFFFAOYNA-N
Synonym 1,4-butanediol diglycidyl ether, 1,4-diglycidyloxybutane, 1,4-bis glycidyloxy butane, butanediol diglycidyl ether, araldit dy 026, 1,4-butane diglycidyl ether, grilonit rv 1806, 1,4-diglycidloxybutane, chs-rr2, 1,4-bis 2,3-epoxypropoxy butane
PubChem CID 17046
IUPAC Name 2-[4-(oxiran-2-ylmethoxy)butoxymethyl]oxirane
SMILES C(CCOCC1CO1)COCC1CO1

Specifications

CAS Min % 60
Color Yellow
Density 1.1000g/mL
Boiling Point 266.0°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 55% min. (GC)
Packaging Glass bottle
Refractive Index 1.4600 to 1.4650
Quantity 500 g
Specific Gravity 1.1
Solubility Information Solubility in water: 55,6g/L (20°C).
Viscosity <15 mPa.s (25°C)
Formula Weight 202.25
Percent Purity 60%
Grade Technical
Physical Form Liquid
Chemical Name or Material 1, 4-Butanediol diglycidyl ether, 60%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Harmful in contact with skin.
Causes serious eye damage.
Harmful if inhaled.
Harmful if swallowed.
Harmful to aquatic life with long l
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
EINECSNumber 219-371-7
RTECSNumber EJ5100000
TSCA TSCA
Recommended Storage This product unavoidably and progressively degrades on storage

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.