Learn More
1,4-Diazabicyclo[2.2.2]octane, 98%
Description
1,4-Diazabicyclo[2.2.2]octane is used as polyurethane catalyst, Balis-Hillman reaction catalyst complexing ligand and lewis base. It finds use in dye lasers and in mounting samples for fluorescence microscopy and as anti-fade reagent shown to scavenge free radicals due to flurochrome excitation of fluorochromes. Further, it is an oxidation and polymerization catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,4-Diazabicyclo[2.2.2]octane is used as polyurethane catalyst, Balis-Hillman reaction catalyst complexing ligand and lewis base. It finds use in dye lasers and in mounting samples for fluorescence microscopy and as anti-fade reagent shown to scavenge free radicals due to flurochrome excitation of fluorochromes. Further, it is an oxidation and polymerization catalyst.
Solubility
Soluble in water, acetone, benzene, ethanol and methyl ethyl ketone.
Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.
Specifications
Specifications
| Melting Point | 155°C to 160°C |
| Density | 1.14 |
| Boiling Point | 174°C to 176°C |
| Flash Point | 62°C (143°F) |
| Odor | Amine-like |
| Quantity | 25 g |
| UN Number | UN1325 |
| Beilstein | 103618 |
| Sensitivity | Hygroscopic |
| Merck Index | 14,9669 |
| Show More |
Safety and Handling
| Hazard Category | H228-H302-H315-H318-H500 |
| Hazard Statement | GHS H Statement H228-H318-H302-H335-H315 Flammable solid. Causes serious eye damage. Harmful if swallowed. May cause respiratory irritation. Causes skin irritation. |
| Precautionary Statement | P210-P240-P241-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P332+P313-P362-P370+P378q-P501c |
| DOTInformation | Hazard Class: 4.1; Packaging Group: II |
| EINECSNumber | 205-999-9 |
| RTECSNumber | HM0354200 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.