Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1,4-Dioxane, 99.5%, Extra Dry over Molecular Sieve, stabilized, AcroSeal™
Click to view available options
Quantity:
500 mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 12.0°C |
| Density | 1.0340g/mL |
| Boiling Point | 101.0°C |
| Quantity | 500 mL |
| Flash Point | 12°C |
| Refractive Index | 1.4205 to 1.4220 |
| Specific Gravity | 1.034 |
| Viscosity | 1.32 mPa.s (20°C) |
| Water | 0.005% max. |
| Solubility Information | Solubility in water: soluble. Other solubilities: soluble in ethanol, acetone, benzene, toluene,, ether and other common organic solvents |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Causes serious eye irritation. Suspected of causing cancer. May cause respiratory irritation. Highly flammable liquid and vapor. May form explosive peroxides. Repeated exposure may cause skin dryness or cracking. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Keep container tightly closed. Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction