Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-(4-Methoxybenzyl)piperazine, 97%
Click to view available options
Quantity:
1 g
5 g
25 g
Description
1-(4-Methoxybenzyl)piperazine is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-(4-Methoxybenzyl)piperazine is used as a pharmaceutical intermediate.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
1-(4-Methoxybenzyl)piperazine is used as a pharmaceutical intermediate.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Specifications
Specifications
Melting Point | 32°C to 36°C |
Color | White to Yellow |
Flash Point | >110°C (230°F) |
Quantity | 5 g |
Synonym | 1-4-methoxybenzyl piperazine, 1-4-methoxyphenyl methyl piperazine, piperazine,1-4-methoxyphenyl methyl, 1-4-methoxy-benzyl-piperazine, piperazine, 1-4-methoxyphenyl methyl, 4-methoxy-1-piperazinylmethyl benzene, 1-4-methoxybenzyl-piperazine, 1-4-methoxybenzyl piperazine #, piperazine, 1-4-methoxybenzyl, 1-4-methoxy-phenylmethyl piperazine |
Formula Weight | 206.29 |
Percent Purity | 97% |
Physical Form | Solid |
Chemical Name or Material | 1-(4-Methoxybenzyl)piperazine |
Safety and Handling
Hazard Category | H302 |
Hazard Statement | GHS H Statement H302 Harmful if swallowed. |
Precautionary Statement | P264b-P270-P301+P312-P330-P501c |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction