Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Pentadiene, 99%

Catalog No. p-178643
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

CAS: 591-93-5 | C5H8 | 68.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-93-5
Molecular Formula C5H8
Molecular Weight (g/mol) 68.11
MDL Number MFCD00008665
InChI Key QYZLKGVUSQXAMU-UHFFFAOYSA-N
Synonym 1,4-pentadiene, unii-a5if861oq5, pentadiene-1,4, allylethylene, penta-1,4-dien-1-yl, penta-1,4-dien-3-yl, ch2=chch2ch=ch2, acmc-1at58
PubChem CID 11587
IUPAC Name penta-1,4-diene
SMILES C=CCC=C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -148.0°C
Color Colorless
Density 0.6500g/mL
Boiling Point 26.0°C
Flash Point −20°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.3884 to 1.3904
Linear Formula H2C=CHCH2CH=CH2
Quantity 1 g
Beilstein 01, 251
Fieser 01,783
Specific Gravity 0.65
Formula Weight 68.11
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1,4-Pentadiene, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be fatal if swallowed and enters airways.
Extremely flammable liquid and vapor.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfac
EINECSNumber 209-736-9
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.