Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5,7-Triazabicyclo[4.4.0]dec-5-ene 98.0+%, TCI America™
SDP

Catalog No. T198225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T198225G 25 g
T19825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T198225G Supplier TCI America Supplier No. T198225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5807-14-7
Molecular Formula C7H13N3
Molecular Weight (g/mol) 139.202
MDL Number MFCD00043003
InChI Key FVKFHMNJTHKMRX-UHFFFAOYSA-N
Synonym 1,3,4,6,7,8-Hexahydro-2H-pyrimido[1,2-a]pyrimidine, TBD
PubChem CID 79873
IUPAC Name 3,4,6,7,8,9-hexahydro-2H-pyrimido[1,2-a]pyrimidine
SMILES C1CNC2=NCCCN2C1

Specifications

Melting Point 130°C
Color White-Yellow
Quantity 25 g
UN Number 3263
Formula Weight 139.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,5,7-Triazabicyclo[4.4.0]dec-5-ene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.