Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,9-Dichlorononane, 98%, Thermo Scientific™

Catalog No. p-7055870
Click to view available options
Quantity:
5 g
25 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as pharmaceutical intermediate. Many organisms use 1,9-dichlorononane as a sole carbon source. Bacteria on incubation of sewage with 1,9-dichlorononane increased its subsequent capacity to dehalogenate.

Solubility
Insoluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 821-99-8
Molecular Formula C9H18Cl2
Molecular Weight (g/mol) 197.143
MDL Number MFCD00001024
InChI Key JMGRNJZUQCEJDB-UHFFFAOYSA-N
Synonym nonane, 1,9-dichloro, 1,9-dichlornonane, nonane,9-dichloro, ksc449k9f, 1,9-dichlornonan;1,9-dichloro-nonan;nonane, 1,9-dichloro-;nonane,1,9-dichloro-;
PubChem CID 13191
IUPAC Name 1,9-dichlorononane
SMILES C(CCCCCl)CCCCCl

Specifications

Color Colorless
Density 1.09 g/mL
Boiling Point 258°C to 262°C
Flash Point >112°C (234°F)
Refractive Index 1.459
Linear Formula Cl(CH2)9Cl
Quantity 5 g
Beilstein 1698729
Solubility Information Insoluble in water.
Formula Weight 197.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1,9-Dichlorononane
Show More Show Less
Hazard Category H315-H319
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 212-484-2
TSCA Yes
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.