Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Acetylpiperidine-4-carbonitrile, 97%, Thermo Scientific™

Catalog No. sew04588ea
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SEW04588EA 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SEW04588EA Supplier Thermo Scientific Chemicals Supplier No. SEW04588EA
Only null left
Add to Cart

Chemical Identifiers

CAS 25503-91-7
Molecular Formula C8H12N2O
Molecular Weight (g/mol) 152.20
MDL Number MFCD00221055
InChI Key NFDGRMQIOHRQHF-UHFFFAOYSA-N
Synonym 1-acetyl-4-piperidinecarbonitrile, 1-acetyl-4-cyanopiperidine, 1-acetyl-isonipecotonitrile, 4-piperidinecarbonitrile,1-acetyl, 1-ethanoylpiperidine-4-carbonitrile
PubChem CID 2756286
IUPAC Name 1-acetylpiperidine-4-carbonitrile
SMILES CC(=O)N1CCC(CC1)C#N

Specifications

Assay Percent Range 97%
Quantity 10 g
Formula Weight 152.2
Chemical Name or Material 1-Acetylpiperidine-4-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.