Learn More
1-Amino-4-cyclopentylpiperazine, 97%
Description
1-Amino-4-cyclopentylpiperazine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Amino-4-cyclopentylpiperazine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Slightly soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Specifications
| Quantity | 1 g |
| Synonym | 1-amino-4-cyclopentylpiperazine, 1-amino-4-cyclopentyl piperazine, 4-cyclopentyl-1-piperazinamine, pubchem8581, 4-cyclopentylpiperazinylamine, 1-piperazinamine,4-cyclopentyl, 1-piperazinamine, 4-cyclopentyl, 4-cyclopentyl-piperazin-1-ylamine, 1-n-cyclopentyl piperazine-4-amine, 1-piperazinamine, 4-cyclopentyl-9ci |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 169.27 |
| Percent Purity | 97% |
| Chemical Name or Material | 1-Amino-4-cyclopentylpiperazine |
Safety and Handling
| Hazard Category | H302-H314-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| EINECSNumber | 262-742-3 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.