Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Aminobenzotriazole, 97%

Catalog No. AAJ6361006
Click to view available options
Quantity:
250 mg
1 g
5 g

Catalog No. AAJ6361006

Supplier: Thermo Scientific Chemicals J6361006

Only null left
Add to Cart

Suicide inhibitor of cytochrome P450 and chloroperoxidase | CAS: 1614-12-6 | C6H6N4 | 134.142 g/mol

1-Aminobenzotriazole is used as a Suicide inhibitor of cytochrome P450 and chloroperoxidase, an inhibitor of oxidative drug metabolism in biochemical pharmacology and toxicology studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Aminobenzotriazole is used as a Suicide inhibitor of cytochrome P450 and chloroperoxidase, an inhibitor of oxidative drug metabolism in biochemical pharmacology and toxicology studies.

Solubility
Soluble in chloroform, DMSO, ethanol or water

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1614-12-6
Molecular Formula C6H6N4
Molecular Weight (g/mol) 134.142
MDL Number MFCD00132902
InChI Key JCXKHYLLVKZPKE-UHFFFAOYSA-N
Synonym 1-aminobenzotriazole, 1h-benzotriazol-1-amine, 1h-benzo d 1,2,3 triazol-1-amine, 1-benzotriazolamine, 1-abtz, 1h-1,2,3-benzotriazol-1-amine, 1h-1,2,3-benzotriazol-1-ylamine, 1,2,3-benzotriazol-1-amine, benzotriazolylamine, 1-benzotriazolylamine
PubChem CID 1367
IUPAC Name benzotriazol-1-amine
SMILES C1=CC=C2C(=C1)N=NN2N

Specifications

Melting Point 81°C to 85°C
Color Brown to White
Quantity 5 g
Beilstein 607843
Solubility Information Soluble in chloroform, DMSO, ethanol or water
Formula Weight 134.14
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1-Aminobenzotriazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber DM1235000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.