Learn More
1-Aminocyclobutanecarboxylic acid, 95%
Description
ACBC is an NMDA receptor antagonist acting at the glycine site. It acts as an agonist being an analog of glycine at the NMDA-glycine receptor site. This affects the signal transmission in the CNS.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ACBC is an NMDA receptor antagonist acting at the glycine site. It acts as an agonist being an analog of glycine at the NMDA-glycine receptor site. This affects the signal transmission in the CNS.
Solubility
Soluble to 100 mM in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Specifications
Specifications
Melting Point | 261°C (decomposition) |
Color | White |
Quantity | 1 g |
Beilstein | 2802253 |
Solubility Information | Soluble to 100 mM in water. |
Formula Weight | 115.13 |
Percent Purity | 95% |
Physical Form | Powder |
Chemical Name or Material | 1-Aminocyclobutanecarboxylic acid |
Safety and Handling
Hazard Category | H302-H315-H319-H335 |
Hazard Statement | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
RTECSNumber | GU1336000 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.