Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzyl-4-cyano-4-hydroxypiperidine, 98%, Thermo Scientific™

Catalog No. AC136860250
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle

Catalog No. AC136860250

Supplier: Thermo Scientific Chemicals 136860250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6094-60-6
Molecular Formula C13H16N2O
Molecular Weight (g/mol) 216.28
MDL Number MFCD00023752
InChI Key XQRLXUYZKZXSBN-UHFFFAOYSA-N
Synonym 1-benzyl-4-cyano-4-hydroxypiperidine, maybridge1_006722, acmc-20aoxx, 1-benzyl-4-hydroxyisonipecotonitrile, 4-hydroxy-1-benzylpiperidine-4-carbonitrile, 1-benyl-4-hydroxy-piperidine-4-carbonitrile, 1-benzyl-4-hydroxy-4-piperidinecarbonitrile, 1-benzyl-4-hydroxy-piperidine-4-carbonitrile, 4-hydroxy-1-phenylmethyl-4-piperidinecarbonitrile
PubChem CID 80189
IUPAC Name 1-benzyl-4-hydroxypiperidine-4-carbonitrile
SMILES OC1(CCN(CC2=CC=CC=C2)CC1)C#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 95°C to 102°C
Color Yellow to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Formula Weight 216.28
Percent Purity ≥97.5%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 1-Benzyl-4-cyano-4-hydroxypiperidine, 98%
Show More Show Less
EINECSNumber 228-043-2
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.