Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-2-chloroethane, 98%, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
Packaging:
Glass bottle
107-04-0
C2H4BrCl
143.41
MFCD00000962
IBYHHJPAARCAIE-UHFFFAOYSA-N
ethylene chlorobromide, 2-chloroethyl bromide, 1-chloro-2-bromoethane, 2-bromoethyl chloride, ethane, 1-bromo-2-chloro, 2-bromo-1-chloroethane, s-chlorobromoethane, 1,2-bromochloroethane, beta-chloroethyl bromide, sym-chlorobromoethane
7849
CHEBI:19032
1-bromo-2-chloroethane
C(CBr)Cl
107-04-0
C2H4BrCl
143.41
MFCD00000962
IBYHHJPAARCAIE-UHFFFAOYSA-N
ethylene chlorobromide, 2-chloroethyl bromide, 1-chloro-2-bromoethane, 2-bromoethyl chloride, ethane, 1-bromo-2-chloro, 2-bromo-1-chloroethane, s-chlorobromoethane, 1,2-bromochloroethane, beta-chloroethyl bromide, sym-chlorobromoethane
7849
CHEBI:19032
1-bromo-2-chloroethane
C(CBr)Cl
Specific Gravity 1.73
Refractive Index 1.4900 to 1.4920
Melting Point -17.0°C
Boiling Point 106.0°C to 107.0°C
Packaging Glass bottle
Density 1.7300g/mL
Infrared Spectrum Authentic
Quantity 500mL
Color Colorless to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula ClCH2CH2Br
Solubility Information Solubility in water: 7g/L (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 143.41
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 1-Bromo-2-chloroethane
Show More Show Less

GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Toxic if inhaled.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove to fresh air

GHS Signal Word: Danger

EINECSNumber : 203-456-