Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-2-fluorobenzene, 99%

Catalog No. AAA1063522
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 1072-85-1 | C6H4BrF | 175 g/mol

1-Bromo-2-fluorobenzene is used as a reactant with cyclopenta-1,3-diene to produce 1,4-dihydro-1,4-methano-naphthalene. This reaction will need reagent Mg, and solvent diethyl ether. The reaction time is 3 hours with the temperature of 20°C,

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-2-fluorobenzene is used as a reactant with cyclopenta-1,3-diene to produce 1,4-dihydro-1,4-methano-naphthalene. This reaction will need reagent Mg, and solvent diethyl ether. The reaction time is 3 hours with the temperature of 20°C,

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from Strong oxidizing agents .
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072-85-1
Molecular Formula C6H4BrF
Molecular Weight (g/mol) 175
MDL Number MFCD00000282
InChI Key IPWBFGUBXWMIPR-UHFFFAOYSA-N
Synonym 2-bromofluorobenzene, o-bromofluorobenzene, 2-fluorobromobenzene, o-fluorobromobenzene, 2-bromfluorbenzen, benzene, 1-bromo-2-fluoro, 1-fluoro-2-bromobenzene, benzene, 1-fluoro-2-bromo, ofbb, benzene, bromofluoro
PubChem CID 61259
IUPAC Name 1-bromo-2-fluorobenzene
SMILES C1=CC=C(C(=C1)F)Br

Specifications

Melting Point -8°C
Density 1.601
Boiling Point 155°C to 157°C
Flash Point 43°C (109°F)
Odor Aromatic
Refractive Index 1.534
Quantity 100 g
UN Number UN1993
Beilstein 1071819
Solubility Information Insoluble in water.
Formula Weight 175.01
Percent Purity 99%
Chemical Name or Material 1-Bromo-2-fluorobenzene
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H302-H319
Flammable liquid and vapor.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 214-018-3
RTECSNumber DA1050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.