Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-4-methoxybutane 98.0+%, TCI America™
SDP

Catalog No. B449525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B449525G 25 g
B44955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B449525G Supplier TCI America Supplier No. B449525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4457-67-4
Molecular Formula C5H11BrO
Molecular Weight (g/mol) 167.046
MDL Number MFCD00037102
InChI Key ALOQTNHQNMYBDE-UHFFFAOYSA-N
Synonym 4-Bromobutyl Methyl Ether
PubChem CID 521168
IUPAC Name 1-bromo-4-methoxybutane
SMILES COCCCCBr

Specifications

Color Colorless
Boiling Point 165°C
Quantity 25 g
UN Number 1993
Formula Weight 167.05
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-4-methoxybutane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.