Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
(1-Butyl)triethylammonium bromide, 97%
Click to view available options
Quantity:
25 g
100 g
Description
(1-Butyl)triethylammonium bromide is an important organic intermediate. It can be used in agrochemical, pharmaceutical & dye stuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(1-Butyl)triethylammonium bromide is an important organic intermediate. It can be used in agrochemical, pharmaceutical & dye stuff field.
Solubility
Soluble in water.
Notes
Hygroscopic. Store away from oxidizing agents, water/ moisture. Store under inert gas.
(1-Butyl)triethylammonium bromide is an important organic intermediate. It can be used in agrochemical, pharmaceutical & dye stuff field.
Solubility
Soluble in water.
Notes
Hygroscopic. Store away from oxidizing agents, water/ moisture. Store under inert gas.
Specifications
Specifications
| Melting Point | 212°C to 215°C |
| Quantity | 25 g |
| Sensitivity | Hygroscopic |
| Solubility Information | Soluble in water. |
| Formula Weight | 238.21 |
| Percent Purity | 97% |
| Chemical Name or Material | (1-Butyl)triethylammonium bromide |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction