Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butyn-3-yl p-Toluenesulfonate 98.0+%, TCI America™
SDP

Catalog No. T12245G
Click to view available options
Quantity:
5 g

Chemical Identifiers

CAS 53487-52-8
Molecular Formula C11H12O3S
Molecular Weight (g/mol) 224.274
MDL Number MFCD00060095
InChI Key XWRDBUBKBUYLQI-UHFFFAOYSA-N
Synonym p-Toluenesulfonic Acid 1-Butyn-3-yl Ester
PubChem CID 561098
IUPAC Name but-3-yn-2-yl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OC(C)C#C

Specifications

Melting Point 52°C
Quantity 5 g
Formula Weight 224.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Butyn-3-yl p-Toluenesulfonate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.