Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Chloro-2,3,3-trifluorocyclobutene, 97%, Thermo Scientific™

Catalog No. AC309670010
Click to view available options
Quantity:
1 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 694-62-2
Molecular Formula C4H2ClF3
Molecular Weight (g/mol) 142.51
MDL Number MFCD00041524
InChI Key NTBUBNMKBVNZBB-UHFFFAOYSA-N
Synonym 1-chloro-2,3,3-trifluorocyclobut-1-ene, cyclobutene,1-chloro-2,3,3-trifluoro, 1-chloro-2,3,3-trifluoro-1-cyclobutene
PubChem CID 136506
IUPAC Name 1-chloro-2,3,3-trifluorocyclobut-1-ene
SMILES FC1=C(Cl)CC1(F)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 142.51
Chemical Name or Material 1-Chloro-2, 3, 3-trifluorocyclobutene, 97%
missing translation for 'healthHazard1' AIR SENSITIVE
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.